missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Yeast Extract (Granulated)
Description
Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins
- Water-soluble portion of autolyzed yeast containing vitamin B complex
- Used in preparing microbiological culture media
- CAS: 8013-01-2
Specifications
Specifications
| Form | Dehydrated |
| Media Type | Yeast Extract |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Used for preparing microbiological culture media |
| Includes | Vitamin B complex |
| Packaging Type | Poly Bottle |
| Quantity | 500 g |