missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Agar, Powder / Flakes, Fisher BioReagents™

Catalog No. BP14232
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP14232 2000 g Poly Pail
BP1423500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP14232 Supplier Fisher BioReagents™ Supplier No. BP14232
Edge

Used as a solidifying agent for culture media.

Used as a solidifying agent for culture media

  • Used as a solidfying agent to provide the solid medium to support the growth of microbes for cell culture applications
  • Functionally tested for promotion of growth of E. coli and S. aureus
  • Typical concentration is 1.5% agar (15g/L) in water
  • CAS: 9002-18-0
  • Molecular Weight: 336.337 g/mol
  • Chemical Formula: C14H24O9

Specifications

Form Powder
Grade Molecular genetics
Product Type Agar
For Use With (Application) Used for a solidifying agent for culture media
Packaging Type Poly Pail
Quantity 2000 g