missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox

Catalog No. p-4907856
Change view
Click to view available options
Quantity:
500 g
2000 g
Packaging Type:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP1427500 500 g Poly Bottle
BP14272 2000 g Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1427500 Supplier Fisher BioReagents™ Supplier No. BP1427500
Only null left
Edge

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
  • Provides a nutrient-rich environment to enable faster growth of E. coli
  • Format: Dehydrated powder

Specifications

Description Bacterial Culture
Form Powder
Grade Molecular genetics
Media Type LB Broth - Lennox
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L
For Use With (Application) Preparing bacterial nutrient media
Packaging Type Poly Bottle
Quantity 500 g