missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Agar, Miller (Powder)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Supplier: Fisher BioReagents™ BP14252
Description
- Directions for Media Preparation: Suspend 40g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
Specifications
| Dehydrated Powder | |
| LB Agar | |
| Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L | |
| Poly Bottle |
| Molecular genetics | |
| Agar | |
| Preparing bacterial nutrient media | |
| 2000 g |