missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media Additives: Yeast Extract

Catalog No. BP1422100
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1422100 100 g
BP1422500 500 g
BP14222 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP1422100 Supplier Fisher BioReagents™ Supplier No. BP1422100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Additive for preparing bacterial nutrient media

Water soluble portion of autolyzed fresh yeast used in bacteriological culture media

Specifications

Form Dehydrated
Grade Molecular genetics
Media Type Yeast Extract
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 100 g