missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Agar, Miller (Granulated)
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Supplier: Fisher BioReagents™ BP97242
Description
- Directions for Media Preparation: Add 32g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
- Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
- Agar provides solid support for growth
- Format: Granulated powder
Specifications
| Granulated Powder | |
| Agar | |
| For maintenance and propagation of Escherichia coli in molecular biology procedures | |
| 2 kg |
| LB Agar, Miller | |
| Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 12g/L | |
| Poly Bottle |