missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ LB Agar, Miller (Granulated)

Catalog No. BP9724500
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP9724500 500 g
BP97242 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP9724500 Supplier Fisher BioReagents™ Supplier No. BP9724 500
Only null left

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 32g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Sodium chloride concentration of Miller formulation is 2X the concentration of the Lennox formulation to allow for optimal salt concentration for a specific strain
  • Agar provides solid support for growth
  • Format: Granulated powder

Specifications

Form Granulated Powder
Media Type LB Agar, Miller
Product Type Nutrient Agar
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 12g/L
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Packaging Type Poly Bottle
Quantity 500 g