missing translation for 'onlineSavingsMsg'
Learn More

Granulated Agar, Fisher BioReagents

Catalog No. BP9744500
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP9744500 500 g Poly Bottle
BP97445 5 kg Poly Pail
BP97442 2 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP9744500 Supplier Thermo Fisher Scientific Supplier No. BP9744500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used as a solidifying agent to provide the solid medium to support the growth of microbes

  • Granulated agar media CAS:[9002-18-0]
  • Store at room temperature

Specifications

Form Granulated Powder
Product Type Agar
For Use With (Application) Used as a solidifying agent to provide the solid medium to support the growth of microbes
Packaging Type Poly Bottle
Quantity 500 g