missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulated Agar, Fisher BioReagents
Description
- Granulated agar media CAS:[9002-18-0]
- Store at room temperature
Spécifications
Spécifications
| Form | Granulated Powder |
| Product Type | Agar |
| For Use With (Application) | Used as a solidifying agent to provide the solid medium to support the growth of microbes |
| Packaging Type | Poly Pail |
| Quantity | 5 kg |