missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Adenosine 5'-triphosphate Disodium Salt Trihydrate

Catalog No. BP41325
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP41325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP41325 Supplier Fisher BioReagents™ Supplier No. BP413 25
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disodium Salt ATP

  • C10H14N5O13P3Na2·3H2O; F.W. 605.24
  • CAS: 51963-61-2
  • Molar Extinction Coefficient (259nm, phosphate buffer, pH 7.0); >15,000
  • Solubility (5% in H2O, 25°C): clear and haze-free
  • Lead: ≤0.002%

Suitable for use in many molecular biology applications

Specifications

Format Solid
pH 7
Molecular Weight (g/mol) 605.24
Content And Storage <0°C
Quantity 25 g
Color White
For Use With (Application) Molecular biology