missing translation for 'onlineSavingsMsg'
Learn More

Fisetin 96.0+%, TCI America™

Catalog No. T01215G
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01215G 5 g
T0121100MG 100 mg
T01211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T01215G Supplier TCI America Supplier No. T01215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 528-48-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.239
MDL Number MFCD00006829
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
Synonym fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
PubChem CID 5281614
ChEBI CHEBI:42567
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O

Specifications

Melting Point 330°C
Quantity 5 g
Formula Weight 286.24
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fisetin
RTECSNumber LK9250000
TSCA Yes