missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fisetin

Catalog No. p-7214831
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC119150010 1 g
AC119150050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC119150010 Supplier Thermo Scientific Chemicals Supplier No. 119150010
Only null left

CAS: 528-48-3 | C15H10O6 | 286.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-48-3
Molecular Formula C15H10O6
Molecular Weight (g/mol) 286.24
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxy-4H-chromen-4-one
SMILES OC1=CC=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Specifications

Melting Point >330.0°C
Quantity 1 g
Merck Index 15, 4119
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: insoluble in ether, benzene
Formula Weight 286.25
Chemical Name or Material Fisetin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber 208-434-4

RUO – Research Use Only