Learn More
Enzo Life Sciences Fis1 polyclonal antibody (100 g)
Supplier: Enzo ALX21010370100
Host: Rabbit. Isotype: IgG. Immunogen: Synthetic peptide surrounding aa 143 of human Fis1. UniProt: Q9Y3D6. Species Reactivity: Human, Mouse, Rat. Applications: WB. Recommended dilutions: Western Blot (1-4 g/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Affinity purified. Formulation: Liquid. In PBS containig 0.5% BSA, 0.01% thimerosal and 30% glycerol.