missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fire Protection High Performance Pipe Marker, White on Red

Catalog No. 18038110
Change view
Click to view available options
Diameter (English):
10 in. or Larger
2.5 to 4.875 in.
8 to 10 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
18038110 2.5 to 4.875 in.
18038109 8 to 10 in.
18038111 10 in. or Larger
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18038110 Supplier Brady™ Supplier No. 5688IISM

Strap-On High Performance Pipe Markers (B-689) are printed plastic sheets with sealed color graphics for use on pipes in harsh environments.

  • Designed for use on rusty, dirty, wet or rough pipes where pressure sensitive adhesive labels aren’t effective.
  • Resistant to UV light exposure, high humidity, high temperatures, abrasion, chemical reagents and physical agitation.
  • Excellent chemical resistance.
  • Durable corner grommets resist tears and rips to keep marker firmly in place using nylon ties.
  • Comes with two large nylon ties.

Specifications

Adhesive Type Non-adhesive
Coating Laminate
Color White on Red
Display Text
Finish Gloss
Format Sheet
Material Coiled Plastic (B-689)
Product Type Pipe Marker
Sterility Non-sterile
Dimensions (L x W) 16 x 12 in.
Length (English) 16 in.
Width (English) 12 in.
Width (Metric) 304.80 mm
Diameter (English) 2.5 to 4.875 in.
Show More Show Less