Learn More
Thermo Scientific™ Fipronil
Supplier: Thermo Scientific™ 465571000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Fipronil | |
| C12H4Cl2F6N4OS | |
| Solubility aq. soln.: insoluble. Other solubilities: soluble in acetone, acetonitrile, insoluble in toluene, hexane | |
| 437.15 |
| 120068-37-3 | |
| 100 g | |
| 437.15 |
Chemical Identifiers
| 120068-37-3 | |
| C12H4Cl2F6N4OS | |
| 437.15 |
Safety and Handling
GHS H Statement Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
RUO â Research Use Only