Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Supplier: Thermo Scientific™ 9404223
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
- Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Chemical Identifiers
| Non-filtered | |
| ISO 9001, ISO 14001 | |
| Thermo Scientific FinnPipette Stepper | |
| FinnTip |
Specifications
| 25 mL | |
| Non-filtered | |
| ISO 9001, ISO 14001 | |
| Thermo Scientific FinnPipette Stepper |
| 500, 1000, 1500, 2000, 2500 μL | |
| Sterile | |
| Individually Wrapped | |
| FinnTip |