missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Stepper Pipette Tips
GREENER_CHOICE

Catalog No. 21377154
Change view
Click to view available options
Volume (Metric):
500 μL
1250 μL
2.5 mL
5 mL
12.5 mL
25 mL
50 mL
Sterility:
Non-sterile
Sterile
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
21377154 5 mL Non-sterile
21377148 500 μL Non-sterile
21377149 500 μL Sterile
21377150 1250 μL Non-sterile
21377151 1250 μL Sterile
21377152 2.5 mL Non-sterile
21377153 2.5 mL Sterile
21377155 5 mL Sterile
21377156 12.5 mL Non-sterile
21377157 12.5 mL Sterile
21377158 25 mL Non-sterile
21377159 25 mL Sterile
21377160 50 mL Non-sterile
21377161 50 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 21377154 Supplier Thermo Scientific™ Supplier No. 9404200
Only null left

Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.

Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.

  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped

Specifications

Volume (Metric) 5 mL
Volume (Metric) Dispensing 100, 200, 300, 400, 500 μL
Filtered Non-filtered
Sterility Non-sterile
Certifications/Compliance ISO 9001, ISO 14001
Format Bulk
For Use With (Equipment) Thermo Scientific FinnPipette Stepper
Product Line FinnTip