missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Finntip™ Stepper Pipette Tips

Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
- Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Specifications
| Volume (Metric) | 500 μL |
| Volume (Metric) Dispensing | 10, 20, 30, 40, 50 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Certifications/Compliance | ISO 9001, ISO 14001 |
| Format | Individually Wrapped |
| For Use With (Equipment) | Thermo Scientific FinnPipette Stepper |
| Product Line | FinnTip |