missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finntip™ Extended Length Pipette Tips
GREENER_CHOICE

Catalog No. 2137743
Change view
Click to view available options
Volume (Metric):
200 μL
1000 μL
Format:
Box
Bulk
Rack
Sterility:
Non-sterile
Sterile
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Format Sterility
2137743 200 μL Box Non-sterile
22028146 200 μL Rack Non-sterile
2137744 200 μL Rack Sterile
21377303 1000 μL Bulk Non-sterile
21377600 1000 μL Rack Non-sterile
21377601 1000 μL Rack Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2137743 Supplier Thermo Scientific™ Supplier No. 9400100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ensure accurate, precise pipetting. These tips are designed using our knowledge of liquid handling, with uniformly smooth, hydrophobic surfaces that prevent liquid retention.

Ensure accurate, precise pipetting. Thermo Scientific™ Finntip™ Pipette Tips are manufactured using high quality raw materials and the latest molding techniques. Finntips are designed using our knowledge of liquid handling. Our modern automated production facilities, and our expertise, enable us to produce uniformly smooth, hydrophobic surfaces on the inside and outside of our tips, to prevent liquid retention.

Key Features:

  • Smooth, uniform inner surface of the tips prevents liquid retention
  • Available as Sterile or Non-sterile option
  • Packaging options: racked or bulk

Proven quality

  • Sterile options certified free of RNase, DNase, DNA, PCR inhibitors, ATP and endotoxin contamination

Tip Range

  • Extended length tips available for volumes 100, 200 and 1000 μL allowing for better reach with specific labware consumables

Immediate traceability

  • Each tip rack has ink-jetted lot and volume information; the convenient color-coded racks make it easy to match the compatible Finnpipette with the corresponding Finntip pipette tip rack insert
  • Color-coding visible in rack packaging options

Note:

Availability may vary by country.

Specifications

Description Finntip 200 Ext, Non-Filtered, Non-sterile,
Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Model Finntip 200 Ext
Length (English) 3 in.
Length (Metric) 7.8cm
Format Box
Color Code Yellow
For Use With (Equipment) Thermo Scientific Finnpipette 200
Product Line FinnTip
Fit for Purpose Certified Quality
Show More Show Less