Learn More
Thermo Scientific™ Finntip™ Flex™ Pipette Tips
GREENER_CHOICE
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
Supplier:  Thermo Scientific™ 94060810
 View more versions of this product
Catalog No. 1438782
Only null left
Description
Description
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Benefits
  • Excellent for single and multichannel pipetting
  • Available in six volume ranges: 0.2-10 μL, 1-200 μL, 5-300 μL, 100-1000 μL, 50-1200 μL and 10mL
  • Sterile tips are supplied in vacuum sealed, sterilized, 10 x 96 rack and they are certified to be free from DNA, DNase, RNase and Endotoxins
  • Save space with refills
Softer Tip
  • The soft tip reduces the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI)
  • It also ensures a better seal with the pipetter module, ensuring higher accuracy and precision
Specifications
Chemical Identifiers
Non-filtered
Thermo Scientific Finnpipette
FinnTip Flex
Specifications
Finntip Flex 1200, Non-Filtered, Non-sterile, Rack
Non-filtered
Turquoise
3.4 in.
Rack
Turquoise
FinnTip Flex
1200 μL
Non-sterile
Finntip Flex 1200
8.7cm
960 tips
Thermo Scientific Finnpipette
Certified Quality
Product Suggestions
Product Suggestions