missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Finnpipette™ Dispenser
GREENER_CHOICE

Catalog No. 14487200
Change view
Click to view available options
Volume (Metric):
0.2 to 1.0 mL
0.4 to 2.0 mL
1.0 to 5.0 mL
10.0 to 60.0 mL
2.0 to 10.0 mL
5.0 to 30.0 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14487200 0.2 to 1.0 mL
14487201 0.4 to 2.0 mL
14487202 1.0 to 5.0 mL
14487203 2.0 to 10.0 mL
14487204 5.0 to 30.0 mL
14487205 10.0 to 60.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14487200 Supplier Thermo Scientific™ Supplier No. 4421120

Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.

Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.

Features:

  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction

Note:

Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.

Specifications

Product Type Analog Bottletop Dispenser
Certifications/Compliance Individual calibration certificate with serial number
Description Range: 0.2-1.0mL, ±6.0µL
Channels 1
Volume (Metric) 0.2 to 1.0 mL
Precision 2 μL

Note: Not for use on hot plates.