missing translation for 'onlineSavingsMsg'
Learn More

Lynn Peavey Fingerprint Ink

Catalog No. S69019A
Change view
Click to view available options
Weight (English):
2 oz.
4 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (English)
S69019A 2 oz.
S69019B 4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S69019A Supplier Lynn Peavey Supplier No. 5569

Quick drying black ink produces excellent ridge detail.

Specifications

Color Black
For Use With (Application) Provides excellent ridge detail
Weight (English) 2 oz.
Product Type Fingerprint Ink