missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mopec Fingernail Clippings Evidence Collection Kit
Description
Construction
- (1) Tamper-evident, pre-sealed kit envelope
- (1) Kit instruction sheet
- (1) Metal fingernail clipper
- (1) Right-hand envelope containing one paper bindle
- (1) Left-hand envelope containing one paper bindle
- (1) Tamper-evident police evidence seal for resealing kit envelope after specimen collection
Dimensions
- Envelope: 5.5 L x 7.5 W in.
Quantity
- 25 per case
Performance
- Everything needed for fingernail collection for comparing with evidence
Specifications
Specifications
| Dimensions (L x W) | 5.5 x 7.5 in. |
| Product Type | Evidence Collection Kit |
| Quantity | 25/Cs. |