missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Fine Vannas Scissors

Catalog No. 22445690
Change view
Click to view available options
Length (English):
3.25 in.
4 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
22445690 4 in.
22445546 3.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22445690 Supplier Mopec™ Supplier No. AA438

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fine Vannas Scissors are designed to maximize visibility and have a long taper ideal for precise cuts.

  • All micro scissors are made from stainless steel

Specifications

Product Type Vannas Micro Scissors
Blade Material Stainless Steel
Blade Style Fine, Straight
Color Code Gray
Color-coded No
Grade Standard
Handle Material Stainless Steel
Length (English) 4 in.
Length (Metric) 10.16 cm
Material Stainless Steel
Plug Type US-style
Size Standard
Color Silver
Power Source Manual
Show More Show Less