missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Fine Pore Cassettes in Tube Packs

Catalog No. B851734GY
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851734GY Gray
B851734BE Beige
B851734BL Blue
B851734GN Green
B851734LA Lavender
B851734LG Light Green
B851734LI Lilac
B851734OR Orange
B851734TN Tan
B851734WH White
B851734YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. B851734GY Supplier Epredia™ Supplier No. B851734GY
Only null left

For use with very small biopsy tissue samples. These cassettes are compatible with all Epredia PrintMate™ Cassette Printers, have a front-edge lid hinge, and attached lids that are available in 12 color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving

Specifications

Pore Style Fine Pore
Quantity 750/Cs.
Closure Included Yes
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Gray
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Cassette