Learn More
Epredia™ Fine Pore Cassettes
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Supplier: Epredia™ B851735GY
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
| Fine Pore | |
| 1000/Cs. | |
| Attached Lid, Front Hinge | |
| Gray |
| Yes | |
| Loose Packed | |
| No | |
| Cassette |