Learn More
Fine Science Tools FINE FORCEPT
Catalog No. NC9889584
Encompass
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumostar/ Length: 11cm
Catalog No. NC9889584 Supplier Fine Science Tools Supplier No. 1129500
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Style: #5/ Tip Shape: Straight/ Tips: Standard/ Tip Dimensions: 0.1 x 0.06mm/ Alloy / Material: Dumostar/ Length: 11cm