missing translation for 'onlineSavingsMsg'
Learn More

Finasteride

Catalog No. AAJ63454MC
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63454MC 100 mg
AAJ63454ME 500 mg
AAJ6345403 1 g
AAJ6345406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ63454MC Supplier Thermo Scientific Chemicals Supplier No. J63454MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of steroid 5-a-reductase | CAS: 98319-26-7 | C23H36N2O2 | 372.553 g/mol

Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.

Notes
Protect from heat. Store away from oxidizing agents.

Chemical Identifiers

CAS 98319-26-7
Molecular Formula C23H36N2O2
Molecular Weight (g/mol) 372.553
MDL Number MFCD00869737
InChI Key DBEPLOCGEIEOCV-WSBQPABSSA-N
Synonym finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
PubChem CID 57363
ChEBI CHEBI:5062
IUPAC Name (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
SMILES CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C

Specifications

Color White
Quantity 100 mg
Merck Index 14,4082
Solubility Information Soluble in water,DMSO,ethanol,methanol,and chloroform.
Formula Weight 372.54
Physical Form Powder
Chemical Name or Material Finasteride
Hazard Category H302-H360D
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
RTECSNumber CL5245000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only