Learn More
Finasteride
Inhibitor of steroid 5-a-reductase | CAS: 98319-26-7 | C23H36N2O2 | 372.553 g/mol
Supplier: Thermo Scientific Chemicals J6345406
| Quantity | 5 g |
|---|
Description
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsFinasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.
Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.
Notes
Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
| 98319-26-7 | |
| 372.553 | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| 57363 | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide |
| C23H36N2O2 | |
| MFCD00869737 | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| CHEBI:5062 | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C |
Specifications
| Finasteride | |
| White | |
| 5 g | |
| 14,4082 | |
| Soluble in water,DMSO,ethanol,methanol,and chloroform. | |
| CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C | |
| 372.553 | |
| CHEBI:5062 | |
| Powder |
| 98319-26-7 | |
| C23H36N2O2 | |
| MFCD00869737 | |
| finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin | |
| DBEPLOCGEIEOCV-WSBQPABSSA-N | |
| (1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide | |
| 57363 | |
| 372.54 |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H360D
RTECSNumber : CL5245000
TSCA : No
Recommended Storage : Store at -20°C