missing translation for 'onlineSavingsMsg'
Learn More

Finasteride

Inhibitor of steroid 5-a-reductase | CAS: 98319-26-7 | C23H36N2O2 | 372.553 g/mol

Supplier:  Thermo Scientific Chemicals J6345403

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6345403


Only null left

Description

Description

Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Finasteride is used as a inhibitor of steroid 5-a-reductase, antiandrogen, an intracellular enzyme that converts the androgen testosterone into 5α dihydrotestosterone.

Solubility
Soluble in water, DMSO, ethanol, methanol, and chloroform.

Notes
Protect from heat. Store away from oxidizing agents.
Specifications

Chemical Identifiers

98319-26-7
372.553
DBEPLOCGEIEOCV-WSBQPABSSA-N
57363
(1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
C23H36N2O2
MFCD00869737
finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
CHEBI:5062
CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C

Specifications

Finasteride
White
1 g
14,4082
Soluble in water,DMSO,ethanol,methanol,and chloroform.
CC12CCC3C(C1CCC2C(=O)NC(C)(C)C)CCC4C3(C=CC(=O)N4)C
372.553
CHEBI:5062
Powder
98319-26-7
C23H36N2O2
MFCD00869737
finasteride, proscar, propecia, finastid, prostide, chibro-proscar, finasterida, finasteridum, finpecia, finasteridum inn-latin
DBEPLOCGEIEOCV-WSBQPABSSA-N
(1S,3aS,3bS,5aR,9aR,9bS,11aS)-N-tert-butyl-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxamide
57363
372.54
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c

H302-H360D

RTECSNumber : CL5245000

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions