missing translation for 'onlineSavingsMsg'
Learn More

Filter aid, Harborlite 800

Catalog No. AAL1360836
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1360836 500 g
AAL136080E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1360836 Supplier Thermo Scientific Chemicals Supplier No. L1360836

CAS: 130885-09-5

Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

Solubility
Insoluble in water.

Notes
Harborlite is a registered trademark of Harborlite Corportion.

Chemical Identifiers

CAS 130885-09-5
MDL Number MFCD00197888
Synonym Perlite, filter aid; Harborlite J2

Specifications

Quantity 500 g
Density 2.3
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Harborlite 800
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber VV7311000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only