missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Replacement filling valves for BRAND™ Bottletop Dispensers
Description
- Filling valves for the Dispensette™ III, Organic, TA and seripettor™ pro are provided with a sealing washer. The updated design of the Dispensette™ S series of bottletop dispensers has eliminated the need for a sealing washer.
- Filling valve with olive-shaped nozzle is recommended for Dispensette™ III and Organic bottletop instruments that are frequently autoclaved.
- seripettor™ valve set includes both the filling and discharge valves for the instrument.
- Calibrating-mounting tool is used for calibrating analog-adjustable Dispensette™ models, as well as removing the valves for routine cleaning or replacement.
Specifications
Specifications
| Product Type | Filling Valve |
| Autoclavable | Autoclavable |
| Color | Translucent/White |
| Material | ETFE, Borosilicate Glass, Ceramic |
| For Use With (Equipment) | Dispensette S or S Organic - 1, 2, 5 and 10 mL |
| Includes | Filling Valve |
| Sterility | Non-sterile |