missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ficoll∣r 400
Description
Ficoll™ 400 is used for cell separation and organelle isolation. It is also used to separate blood to its components like erythrocytes and leukocytes. It acts as a mononuclear cell separator and used in culture media, nucleic acidhybridization, electrophoresis and in immunological studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 50 g |
| Solubility Information | Soluble in water. |
| Formula Weight | ca 400,000 |
| Chemical Name or Material | Ficoll 400 |
Safety and Handling
| EINECSNumber | 200-334-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only