missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ficoll PM70 Perfusion Media

Catalog No. 45002019
Change view
Click to view available options
Product Type:
Perfusion Media
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
45002019 Perfusion Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002019 Supplier Cytiva Supplier No. 17031050
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A high molecular weight sucrose polymer with minimal toxicity and good solubility in aqueous solutions.

  • Stable in neutral and alkaline solutions
  • Rapidly hydrolyzed in pH 3 solutions, especially at high temperature
  • Readily soluble in aqueous media in powder form
  • Good viscosity and osmotic properties
  • 50% concentrations (weight/volume) can be easily achieved

Specifically intended for use in perfusion media for laboratory studies on dextran-sensitive animals

Specifications

Color White
Content And Storage 4° to 30°C
Product Type Perfusion Media
Quantity 0.5 kg
For Use With (Application) Lab study on Dextran-sensitive animals