missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Ficoll PM400

Catalog No. 45001745
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45001745 100 g
45001746 500 g
45001792 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45001745 Supplier Cytiva Supplier No. 17030010
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions

High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. Cytiva™ Ficoll PM400 is a synthetic neutral, highly-branched hydrophilic polymer of sucrose, used for producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity.

  • For producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity
  • Neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution
  • Concentrations of up to 50% (w/v) covering densities of up to 1.2g/mL can be achieved
  • Has better osmotic properties than sucrose
  • Preserves functional and morphological integrity
  • Useful for separating cells that are sensitive to centrifugation and for separating cells of similar density but different size
  • Does not penetrate biological membranes
  • Serves as the raw material for preparation of Ficoll-Paque gradients
  • Used in other applications such as electrophoresis, hybridization, cryopreservation, and as a hapten carrier
  • Supplied as dry powders
  • Used in the formation of density gradients for the separation and isolation of eukaryotic cells, organelles, and bacterial cells, as a stabilizing agent and as a separation medium for the isolation of lymphocytes
  • Used in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies
  • Used for gradient centrifugation in all types of centrifuge rotors and for separation at unit gravity

Specifications

Quantity 100 g
Content And Storage 4–30°C
For Use With (Application) Cell Isolation
Viscosity 5 ηr at 20°C: 10% (w/v), 20 ηr at 20°C: 20% (w/v), 60 ηr at 20°C : 30% (w/v, 180 ηr at 20°C : 40% (w/v) , 600 ηr at 20°C : 50% (w/v)
Endotoxin Level 0.12 EU/mg
Color White
Product Type Centrifugation Media
Density 1.2 g/mL
Form Powder