missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ficoll PM 70 Centrifugation Media
Description
- High molecular weight (70,000) sucrose-polymer formed by copolymerization of sucrose with epichlorohydrin
- Highly branched structure and high content of hydroxyl groups mean good solubility in aqueous media with concentrations up to 50% w/v easily attained
- Displays minimal toxicity, plus favorable viscosity and osmotic properties
- Low endotoxin levels (<0.12 EU/mg)
For use in perfusion media for laboratory studies on dextran-sensitive animals
Specifications
Specifications
| Quantity | 100 g |
| For Use With (Application) | Specifically intended for use in perfusion media for laboratory studies on dextran-sensitive animals. |
| Viscosity | ≈10mL/g |
| Endotoxin Level | 0.12 EU/mg |
| Color | White |
| Product Type | Centrifugation Media |
| Form | Powder |
| Content And Storage | 4–30°C |