missing translation for 'onlineSavingsMsg'
Learn More

Ficoll™, MP Biomedicals™

Numéro de catalogue. ICN16000305
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
ICN16000305 5 g
ICN16000325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. ICN16000305 Fournisseur MP Biomedicals Inc Code fournisseur 0216000305
Il en reste null

Identifiants chimiques

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Spécifications

Color White
Physical Form Powder
Density <1.2 g/mL
Quantity 5 g
Formula Weight ≈400,000
Chemical Name or Material Ficoll™