missing translation for 'onlineSavingsMsg'
Learn More

Ficin from Fig Tree Latex, TCI America™

Catalog No. F001025G
Change view
Click to view available options
Quantity:
25 g
Catalog No. Quantity
F001025G 25 g
1 options

Catalog No. F001025G

Supplier: TCI America F001025G

Only null left

Chemical Identifiers

CAS 9001-33-6
MDL Number MFCD00081597

Specifications

Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Ficin from Fig Tree Latex
RTECSNumber LK3675000
TSCA Yes
Recommended Storage Refrigerator