missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Fibre-Metal™ Face Shield Headgear

Catalog No. 19085619
Change view
Click to view available options
Includes:
Faceshield Adaptor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19085619 Faceshield Adaptor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19085619 Supplier Honeywell™ Supplier No. F5500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for secure, long-term combination use

  • Window is not included and sold separately
  • Quick-Lok™ Cups can be can be securely mounted onto, and removed from the cap without tools

Specifications

Color Gray
Product Type Faceshield Adaptor
For Use With (Equipment) Front Brim Hardhats
Includes Faceshield Adaptor
Material Noryl