missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed Angle Rotor

Catalog No. 15700712
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
15700712 Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15700712 Supplier Thermo Scientific™ Supplier No. 78500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Process large volumes with the six-place, 250 mL fixed angle rotor.

Process large volumes with the six-place, 250mL Thermo Scientific™ Fiberlite™ F14-6x250y Fixed Angle Rotor.

Features:

Fiberlite rotors:

  • Run times are reduced with higher G-forces achieved through lightweight carbon fiber rotors
  • Corrosion- and fatigue-resistant design ensures structural integrity
  • Rotor is backed by a 15-year warranty

Compatible with:

Thermo Scientific™ Sorvall™ RC-6 Plus, Sorvall Evolution™ RC Series, Thermo Scientific™ Jouan KR22i, Beckman™ J2 Series, Beckman J2, Avanti Series, Hitachi™ Himac CR21/CR22 Series, Nalgene™ bottles and tubes

Recommended for:

Processing lysates, homogenates, ammonium sulphate precipitation of proteins and enzymes and plant tissue homogenates

†Performance may vary by centrifuge model.

Specifications

Description Fiberlite F14-6 x 250y Fixed Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 6 x 250 mL
Max. RCF 30,100 x g
Max. Speed 14,000 rpm
Radius (Metric) 137 mm
Radius (Metric) Min. 37 mm
K Factor 1690
Max. Tube Length 135 mm
Max. Tube Diameter 62 mm
Net Weight 15.5 lb. (7.03 kg)
For Use With (Equipment) Thermo Scientific Sorvall RC 6 Plus and Evolution RC Superspeed Centrifuges
Angle 23°
Min. RCF 8,122 x g
Biocontainment Yes
Show More Show Less