missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fiberglass Warning Sign: BIOHAZARD AUTHORIZED PERSONNEL ONLY

Improve workplace safety with proper signage. The Brady™ Warning Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

Specifications

Finish Not Laminated
Material Fiberglass
Quantity 1 Ea.
Adhesive Type Non-adhesive
Mounting Type Corner Holes
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19107321
View Documents Promotion Details
Brady™
62550
10 x 7 in. N/A
Only null left
 
19107326
View Documents Promotion Details
Brady™
BRD62812
14 x 10 in. N/A
Only null left
 
Description

Description

  • Permanently embedded graphics within the sign panel and excellent outdoor durability
  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
Specifications

Specifications

Not Laminated
1 Ea.
Corner Holes
-40°F
Rectangular
BIOHAZARD AUTHORIZED PERSONNEL ONLY
Fiberglass
Non-adhesive
194°F
0.0984 in.
-40°F to 194°F
Black on Orange
Videos
SDS
Documents

Documents