missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Fiberglass Sign: THIS EQUIPMENT STARTS AUTOMATICALLY. STAY CLEAR
Improve workplace safety with proper signage and markings. This Brady™ fiberglass sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$25.67 - $42.97
Specifications
| Material | Fiberglass |
|---|---|
| Shape | Rectangular |
| Displayed Text | THIS EQUIPMENT STARTS AUTOMATICALLY. STAY CLEAR |
| Color | Black on White |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity | |||||
|
19110813
|
Brady™
144068 |
14 x 10 in. |
Each for $42.97
|
|
|||||
|
19-110-818
|
Brady™
144074 |
10 x 7 in. |
Each for $25.67
|
|
|||||
Description
- Fiberglass material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
| Fiberglass | |
| Rectangular | |
| THIS EQUIPMENT STARTS AUTOMATICALLY. STAY CLEAR | |
| Black on White |