missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Fiberglass Danger Sign: HAZARDOUS WASTE STORAGE

Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

$26.07 - $43.64

Specifications

Finish Not Laminated
Material Fiberglass
Quantity 1 Ea.
Adhesive Type Non-adhesive
Mounting Type Corner Holes
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19106182
View Documents Promotion Details
Brady™
131761
10 x 7 in.
Each for $26.07
Only null left
 
19106183
View Documents Promotion Details
Brady™
131762
14 x 10 in.
Each for $43.64
Only null left
 
Description

Description

  • Permanently embedded graphics within the sign panel and excellent outdoor durability
  • Fiberglass material is abrasion-resistant and chemical-resistant
  • Fiberglass out-performs all other sign materials in terms of strength, durability, and longevity
  • Bold and bright, providing high visibility to critical messages
Specifications

Specifications

Not Laminated
1 Ea.
Corner Holes
-40°F
Rectangular
HAZARDOUS WASTE STORAGE
Fiberglass
Non-adhesive
194°F
0.0984 in.
-40°F to 194°F
Red on White
Videos
SDS
Documents

Documents