Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Catalog No. p-4530842
Change view
Click to view available options
Holds:
100 (10 x 10 Grid)
16 (4 x 4 Grid)
25 (5 x 5 Grid)
49 (7 x 7 Grid)
64 (8 x 8 Grid)
81 (9 x 9 Grid)
Quantity:
12 Pack
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds Quantity
1167824C 100 (10 x 10 Grid) Each
1167826D 49 (7 x 7 Grid) 12 Pack
1399233 64 (8 x 8 Grid) 12 Pack
13989218 81 (9 x 9 Grid) 12 Pack
15465468 16 (4 x 4 Grid) Each
13994085 25 (5 x 5 Grid) Each
15465466 49 (7 x 7 Grid) Each
15465465 64 (8 x 8 Grid) Each
15465464 81 (9 x 9 Grid) Each
15465463 100 (10 x 10 Grid) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 1167824C Supplier Thermo Scientific™ Supplier No. 5958
Organize vial storage within cryo boxes with these fiberboard box dividers.
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Quantity Each
Holds 100 (10 x 10 Grid)