Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
- Use with fiberboard storage boxes to divide box into grids for vial storage
- Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Specifications
| Quantity | Each |
| Holds | 81 (9 x 9 Grid) |