Learn More
Lonza Walkersville Inc FGM-3 Cardiac Fibroblast Growth Medium-3 BulletKit
Catalog No. NC0089421
Encompass
FGM-3 Fibroblast Growth Medium-3 was developed specifically for culturing primary cardiac fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-3 Medium contains contains 10% serum.Content:1 x FBM Basal Medium (CC-3131), 500 mL1 x FGM-3 SingleQuots Supplement Pack (CC-4525) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 50.00 mL
Catalog No. NC0089421 Supplier Lonza Walkersville Inc Supplier No. CC4526
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
FGM-3 Fibroblast Growth Medium-3 was developed specifically for culturing primary cardiac fibroblasts. The kit contains basal medium and supplements packaged together for convenience to culture fibroblasts. FGM-3 Medium contains contains 10% serum.Content:1 x FBM Basal Medium (CC-3131), 500 mL1 x FGM-3 SingleQuots Supplement Pack (CC-4525) containing:1 x Lilac Cap Vial with Insulin, 0.50 mL1 x Gray Cap Vial with hFGF-B, 0.50 mL1 x Red Cap Vial with GA-1000, 0.50 mL1 x Bottle FBS, 50.00 mL