missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fexofenadine hydrochloride

Catalog No. AC464970010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464970010 1 g
AC464972500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464970010 Supplier Thermo Scientific™ Supplier No. 464970010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 153439-40-8
Molecular Formula C32H40ClNO4
Molecular Weight (g/mol) 538.13
MDL Number MFCD00865710
InChI Key RRJFVPUCXDGFJB-UHFFFAOYNA-N
IUPAC Name hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
SMILES [H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Formula Weight 538.12
Chemical Name or Material Fexofenadine hydrochloride

RUO – Research Use Only