missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid, Powder, Spectrum™ Chemical

Catalog No. 18610591
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
20 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610591 20 kg Amber Glass Bottle
18610593 5 g Amber Glass Bottle
18610592 25 g Amber Glass Bottle
18610590 100 g Amber Glass Bottle
18610589 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18610591 Supplier Spectrum Chemical Mfg Cor Supplier No. F110620KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1135-24-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 20 kg
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Ferulic Acid, Powder

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.