Learn More
Ferrous Fumarate, USP, 97-101%, Spectrum™ Chemical
FE108, 141-01-5, C4H2FeO4
Supplier:  Spectrum Chemical Mfg Cor FE10812KG
 View more versions of this product
Catalog No. 18602697
Only null left
Explore available promotions
Description
Description
Spectrum™ Chemical Ferrous Fumarate, USP is used primarily to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
141-01-5
12 kg
PMVSDNDAUGGCCE-ODZAUARKSA-L
λ²-iron(2+) (2Z)-but-2-enedioate
97 to 101%
1.5%
100%
C4H2FeO4
[Fe++].[O-]C(=O)\C=C/C([O-])=O
169.90
USP
Poly Pail
Chemical Identifiers
141-01-5
169.90
λ²-iron(2+) (2Z)-but-2-enedioate
C4H2FeO4
PMVSDNDAUGGCCE-ODZAUARKSA-L
[Fe++].[O-]C(=O)\C=C/C([O-])=O
Product Suggestions
Product Suggestions