missing translation for 'onlineSavingsMsg'
Learn More

Ferrous Fumarate, USP, 97-101%, Spectrum™ Chemical

Catalog No. 18602695
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602695 500 g Amber Glass Bottle
18602696 2.5 kg Amber Glass Bottle
18602697 12 kg Poly Pail
18602698 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602695 Supplier Spectrum Chemical Mfg Cor Supplier No. FE108500GM

FE108, 141-01-5, C4H2FeO4

Spectrum™ Chemical Ferrous Fumarate, USP is used primarily to treat iron deficiency anemia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 141-01-5
Molecular Formula C4H2FeO4
Molecular Weight (g/mol) 169.90
InChI Key PMVSDNDAUGGCCE-ODZAUARKSA-L
IUPAC Name λ²-iron(2+) (2Z)-but-2-enedioate
SMILES [Fe++].[O-]C(=O)\C=C/C([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 97 to 101%
Grade USP
Loss on Drying 1.5%
Packaging Amber Glass Bottle